Product Details

SNP ID
hCV89937536
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:26582826 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCACCCCCTCCAGAATGCCTTACT[A/T]CATGAAATCTGGAGACTTGGCCATT
Phenotype
MIM: 610646
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
C2orf70 PubMed Links

Gene Details

Gene
C2orf70
Gene Name
chromosome 2 open reading frame 70
There are no transcripts associated with this gene.

Gene
CIB4
Gene Name
calcium and integrin binding family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029881.2 580 Missense Mutation AAC,TAC N176Y NP_001025052.1

View Full Product Details