Product Details

SNP ID
rs58194358
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:12737714 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACACCAAGTTCCTTACCATCTTT[C/T]TTTATATTCAGTCTTCTTAACTGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM40 PubMed Links

Gene Details

Gene
TMEM40
Gene Name
transmembrane protein 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284406.1 680 Silent Mutation AAA,AAG K171K NP_001271335.1
NM_001284407.1 680 Silent Mutation AAA,AAG K125K NP_001271336.1
NM_001284408.1 680 Silent Mutation AAA,AAG K79K NP_001271337.1
NM_018306.3 680 Silent Mutation AAA,AAG K155K NP_060776.2
XM_011533937.2 680 Silent Mutation AAA,AAG K155K XP_011532239.1

View Full Product Details