Product Details

SNP ID
rs56733391
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:33119942 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTAAGGGACATAGCAGCCCAAGA[A/G]GACTGTGCAGGGAGGGAACCAGGAA
Phenotype
MIM: 605497
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRTAP PubMed Links

Gene Details

Gene
CRTAP
Gene Name
cartilage associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006371.4 Intron NP_006362.1

View Full Product Details