Product Details
- SNP ID
-
hCV90005757
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:45030837 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCCTCCTGCTTCAGATGTTGTGAA[A/C]ACAAAGATGTTTGAGGAGCTCAAGA
- Phenotype
-
MIM: 187520
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CLEC3B
PubMed Links
Gene Details
- Gene
- CLEC3B
- Gene Name
- C-type lectin domain family 3 member B
There are no transcripts associated with this gene.
View Full Product Details