Product Details

SNP ID
rs59554134
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136882018 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAGCTCTCGGTGTGAGCAAGTGG[A/G]CTGCGGCTATTTGAGGTGGGCACTT
Phenotype
MIM: 601895
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR4479 PubMed Links

Gene Details

Gene
MIR4479
Gene Name
microRNA 4479
There are no transcripts associated with this gene.

Gene
TRAF2
Gene Name
TNF receptor associated factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021138.3 243 Intron NP_066961.2
XM_011518973.2 243 Missense Mutation GAC,GGC D16G XP_011517275.1
XM_011518974.2 243 Intron XP_011517276.1
XM_011518975.2 243 Intron XP_011517277.1
XM_011518976.2 243 UTR 5 XP_011517278.1
XM_011518977.2 243 Intron XP_011517279.1
XM_011518978.2 243 Intron XP_011517280.1
XM_017015094.1 243 Intron XP_016870583.1
XM_017015095.1 243 Intron XP_016870584.1

View Full Product Details