Product Details
- SNP ID
-
rs56142402
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:58671560 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTCCGCCCCTGCCTCCGCGGCTCG[A/G]AGGCGAGCGGAAGGTGCCCCGGGGC
- Phenotype
-
MIM: 604860
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC101927322
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs373689542] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC101927322
- Gene Name
- uncharacterized LOC101927322
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001289967.1 |
314 |
Missense Mutation |
TCC,TTC |
S29F |
NP_001276896.1 |
- Gene
- LOC102724745
- Gene Name
- uncharacterized LOC102724745
There are no transcripts associated with this gene.
- Gene
- MALT1
- Gene Name
- MALT1 paracaspase
View Full Product Details