Product Details

SNP ID
rs55779481
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:118815355 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTTAACTTTCCTGGACGCTTCAC[C/T]GGCAAGGCTGCGTCTCTTCTTGTTC
Phenotype
MIM: 610098
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MCM9 PubMed Links

Gene Details

Gene
MCM9
Gene Name
minichromosome maintenance 9 homologous recombination repair factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017696.2 2916 Silent Mutation CCA,CCG P967P NP_060166.2
NM_153255.4 2916 Intron NP_694987.1

View Full Product Details