Product Details
- SNP ID
-
rs56300302
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:121080850 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTTTGTTCCAAAATTTCCATGTAT[G/T]CAAAATAATCACTCACTCGAAACCC
- Phenotype
-
MIM: 615867
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TBC1D32
PubMed Links
Gene Details
- Gene
- TBC1D32
- Gene Name
- TBC1 domain family member 32
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152730.5 |
3947 |
Missense Mutation |
GAA,GCA |
E1232A |
NP_689943.4 |
XM_005266861.2 |
3947 |
Missense Mutation |
GAA,GCA |
E1273A |
XP_005266918.1 |
XM_011535569.1 |
3947 |
Missense Mutation |
GAA,GCA |
E1294A |
XP_011533871.1 |
XM_011535570.1 |
3947 |
Missense Mutation |
GAA,GCA |
E1294A |
XP_011533872.1 |
XM_011535571.2 |
3947 |
Missense Mutation |
GAA,GCA |
E1294A |
XP_011533873.1 |
XM_011535574.1 |
3947 |
Missense Mutation |
GAA,GCA |
E1185A |
XP_011533876.1 |
XM_011535575.2 |
3947 |
Intron |
|
|
XP_011533877.1 |
XM_011535576.2 |
3947 |
Intron |
|
|
XP_011533878.1 |
XM_011535580.2 |
3947 |
Missense Mutation |
GAA,GCA |
E901A |
XP_011533882.1 |
XM_011535582.2 |
3947 |
Intron |
|
|
XP_011533884.1 |
XM_011535585.2 |
3947 |
Intron |
|
|
XP_011533887.1 |
XM_017010397.1 |
3947 |
Missense Mutation |
GAA,GCA |
E1273A |
XP_016865886.1 |
XM_017010398.1 |
3947 |
Missense Mutation |
GAA,GCA |
E1273A |
XP_016865887.1 |
XM_017010399.1 |
3947 |
Missense Mutation |
GAA,GCA |
E1253A |
XP_016865888.1 |
XM_017010400.1 |
3947 |
Missense Mutation |
GAA,GCA |
E1252A |
XP_016865889.1 |
XM_017010401.1 |
3947 |
Missense Mutation |
GAA,GCA |
E1232A |
XP_016865890.1 |
XM_017010402.1 |
3947 |
Intron |
|
|
XP_016865891.1 |
XM_017010403.1 |
3947 |
Intron |
|
|
XP_016865892.1 |
XM_017010404.1 |
3947 |
Missense Mutation |
GAA,GCA |
E690A |
XP_016865893.1 |
XM_017010405.1 |
3947 |
Intron |
|
|
XP_016865894.1 |
View Full Product Details