Product Details

SNP ID
rs59107957
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:96020381 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATGCTGATTCAATATTTATGTCA[C/T]CACAGAGAAGGTTTCGTATCCATAT
Phenotype
MIM: 606865
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FUT9 PubMed Links

Gene Details

Gene
FUT9
Gene Name
fucosyltransferase 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006581.3 Intron NP_006572.2
XM_011535383.2 Intron XP_011533685.1
XM_011535385.2 Intron XP_011533687.1
XM_017010188.1 Intron XP_016865677.1
XM_017010190.1 Intron XP_016865679.1

View Full Product Details