Product Details

SNP ID
rs60242081
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:64746529 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATCCTCCTCTGGATTCCTGCCCC[C/G]CTCCCATGAAGGGAGGAAGAGGATT
Phenotype
MIM: 182870
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PLEKHG3 PubMed Links

Gene Details

Gene
PLEKHG3
Gene Name
pleckstrin homology and RhoGEF domain containing G3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308147.1 10033 Intron NP_001295076.1
Gene
SPTB
Gene Name
spectrin beta, erythrocytic
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000347.5 10033 Intron NP_000338.3
NM_001024858.2 10033 UTR 3 NP_001020029.1
XM_005268023.4 10033 UTR 3 XP_005268080.1
XM_011537105.2 10033 Intron XP_011535407.1
XM_017021612.1 10033 UTR 3 XP_016877101.1
XM_017021613.1 10033 Intron XP_016877102.1
XM_017021614.1 10033 Intron XP_016877103.1

View Full Product Details