Product Details

SNP ID
rs56103835
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:101056219 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATGCTGCGAGCAGTGCCACCTCA[C/T]GGTACTCGGAGGGAGGTTGTCCGTG
Phenotype
MIM: 610164 MIM: 611794 MIM: 614057 MIM: 615036 MIM: 615385 MIM: 615037
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR134 PubMed Links

Gene Details

Gene
MIR134
Gene Name
microRNA 134
There are no transcripts associated with this gene.

Gene
MIR154
Gene Name
microRNA 154
There are no transcripts associated with this gene.

Gene
MIR323B
Gene Name
microRNA 323b
There are no transcripts associated with this gene.

Gene
MIR369
Gene Name
microRNA 369
There are no transcripts associated with this gene.

Gene
MIR377
Gene Name
microRNA 377
There are no transcripts associated with this gene.

Gene
MIR381HG
Gene Name
MIR381 host gene
There are no transcripts associated with this gene.

Gene
MIR382
Gene Name
microRNA 382
There are no transcripts associated with this gene.

Gene
MIR409
Gene Name
microRNA 409
There are no transcripts associated with this gene.

Gene
MIR410
Gene Name
microRNA 410
There are no transcripts associated with this gene.

Gene
MIR412
Gene Name
microRNA 412
There are no transcripts associated with this gene.

Gene
MIR485
Gene Name
microRNA 485
There are no transcripts associated with this gene.

Gene
MIR487A
Gene Name
microRNA 487a
There are no transcripts associated with this gene.

Gene
MIR487B
Gene Name
microRNA 487b
There are no transcripts associated with this gene.

Gene
MIR496
Gene Name
microRNA 496
There are no transcripts associated with this gene.

Gene
MIR539
Gene Name
microRNA 539
There are no transcripts associated with this gene.

Gene
MIR541
Gene Name
microRNA 541
There are no transcripts associated with this gene.

Gene
MIR544A
Gene Name
microRNA 544a
There are no transcripts associated with this gene.

Gene
MIR655
Gene Name
microRNA 655
There are no transcripts associated with this gene.

Gene
MIR668
Gene Name
microRNA 668
There are no transcripts associated with this gene.

Gene
MIR889
Gene Name
microRNA 889
There are no transcripts associated with this gene.

View Full Product Details