Product Details
- SNP ID
-
rs59124487
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:31976995 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAGGTCTATGGTGGGTTCTTGCAA[A/C]ATGAAGCCAGTTGAAGCTGGCATTT
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
XKR7
PubMed Links
Gene Details
- Gene
- XKR7
- Gene Name
- XK related 7
View Full Product Details