Product Details
- SNP ID
-
rs61754172
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:56714614 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAGAATACCCACCTGGGCTTGTTG[G/T]GTGGTTGCCTGGGTGGAATCCTGTT
- Phenotype
-
MIM: 601234
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NACA
PubMed Links
Gene Details
- Gene
- NACA
- Gene Name
- nascent polypeptide-associated complex alpha subunit
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001113201.2 |
426 |
Silent Mutation |
ACA,ACC |
T48T |
NP_001106672.1 |
NM_001113202.1 |
426 |
Silent Mutation |
ACA,ACC |
T48T |
NP_001106673.1 |
NM_001113203.2 |
426 |
Silent Mutation |
ACA,ACC |
T758T |
NP_001106674.2 |
NM_001320193.1 |
426 |
Intron |
|
|
NP_001307122.1 |
NM_001320194.1 |
426 |
Silent Mutation |
ACA,ACC |
T48T |
NP_001307123.1 |
NM_005594.5 |
426 |
Silent Mutation |
ACA,ACC |
T48T |
NP_005585.1 |
XM_006719412.1 |
426 |
Silent Mutation |
ACA,ACC |
T1911T |
XP_006719475.1 |
XM_006719413.2 |
426 |
Silent Mutation |
ACA,ACC |
T1911T |
XP_006719476.1 |
XM_006719414.1 |
426 |
Silent Mutation |
ACA,ACC |
T1911T |
XP_006719477.1 |
XM_006719418.1 |
426 |
Intron |
|
|
XP_006719481.1 |
XM_006719420.3 |
426 |
Silent Mutation |
ACA,ACC |
T48T |
XP_006719483.1 |
XM_011538389.1 |
426 |
Silent Mutation |
ACA,ACC |
T1911T |
XP_011536691.1 |
XM_017019337.1 |
426 |
Intron |
|
|
XP_016874826.1 |
View Full Product Details