Product Details

SNP ID
rs56078738
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56953171 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCATGGTGGCATTCCCTCTGTAT[A/G]GAGTTTCCATATTTTACACACCTGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RDH16 PubMed Links

Gene Details

Gene
RDH16
Gene Name
retinol dehydrogenase 16 (all-trans)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320108.1 Intron NP_001307037.1
NM_003708.4 Intron NP_003699.3

View Full Product Details