Product Details

SNP ID
rs55783970
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:75399964 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAGAAGGAAATGGTATCCCTAAA[C/T]GGTTATTCACTGTACCATCATTTAA
Phenotype
MIM: 607724 MIM: 610394
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CAPS2 PubMed Links

Gene Details

Gene
CAPS2
Gene Name
calcyphosine 2
There are no transcripts associated with this gene.

Gene
GLIPR1L2
Gene Name
GLI pathogenesis-related 1 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270396.1 Intron NP_001257325.1
NM_152436.2 Intron NP_689649.1
XM_011537949.2 Intron XP_011536251.1
XM_011537950.2 Intron XP_011536252.1
XM_017018848.1 Intron XP_016874337.1
XM_017018849.1 Intron XP_016874338.1

View Full Product Details