Product Details

SNP ID
rs56875008
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:110898945 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGGAAAAAAATGCAGGTATCAC[C/T]TTACTCCATTGTTATCTGACCTAGA
Phenotype
MIM: 613605 MIM: 608610
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BBIP1 PubMed Links

Gene Details

Gene
BBIP1
Gene Name
BBSome interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195304.1 1921 UTR 3 NP_001182233.1
NM_001195305.1 1921 UTR 3 NP_001182234.1
NM_001195306.1 1921 UTR 3 NP_001182235.1
NM_001195307.1 1921 UTR 3 NP_001182236.1
NM_001243783.1 1921 UTR 3 NP_001230712.1
Gene
MIR4680
Gene Name
microRNA 4680
There are no transcripts associated with this gene.

Gene
PDCD4
Gene Name
programmed cell death 4 (neoplastic transformation inhibitor)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199492.1 1921 UTR 3 NP_001186421.1
NM_014456.4 1921 UTR 3 NP_055271.2
NM_145341.3 1921 UTR 3 NP_663314.1

View Full Product Details