Product Details

SNP ID
rs60692462
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:122911247 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTCTTAGTTTCATAAAATAATTT[C/T]AAACCTAATATACAGTGAGGGGTGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM24A PubMed Links

Gene Details

Gene
FAM24A
Gene Name
family with sequence similarity 24 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029888.1 Intron NP_001025059.1
XM_017015638.1 Intron XP_016871127.1
XM_017015639.1 Intron XP_016871128.1

View Full Product Details