Product Details

SNP ID
rs61782130
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:51624741 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGAATACTTTAGACAGTCTTGA[C/T]TTTTCCCTTTTCTAAAACTCCTGAG
Phenotype
MIM: 606737
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OSBPL9 PubMed Links

Gene Details

Gene
OSBPL9
Gene Name
oxysterol binding protein like 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024586.5 Intron NP_078862.4
NM_148904.3 Intron NP_683702.1
NM_148905.3 Intron NP_683703.1
NM_148906.2 Intron NP_683704.2
NM_148907.2 Intron NP_683705.1
NM_148908.3 Intron NP_683706.3
NM_148909.3 Intron NP_683707.3
XM_006710318.2 Intron XP_006710381.1
XM_006710320.3 Intron XP_006710383.1
XM_006710321.2 Intron XP_006710384.1
XM_006710322.3 Intron XP_006710385.1
XM_006710323.2 Intron XP_006710386.1
XM_006710324.3 Intron XP_006710387.1
XM_006710325.3 Intron XP_006710388.1
XM_006710326.2 Intron XP_006710389.1
XM_011540599.2 Intron XP_011538901.1
XM_011540600.2 Intron XP_011538902.1
XM_011540601.2 Intron XP_011538903.1
XM_011540602.2 Intron XP_011538904.1
XM_011540603.2 Intron XP_011538905.1
XM_011540604.2 Intron XP_011538906.1
XM_017000218.1 Intron XP_016855707.1
XM_017000219.1 Intron XP_016855708.1
XM_017000220.1 Intron XP_016855709.1
XM_017000221.1 Intron XP_016855710.1
XM_017000222.1 Intron XP_016855711.1
XM_017000223.1 Intron XP_016855712.1
XM_017000224.1 Intron XP_016855713.1
XM_017000225.1 Intron XP_016855714.1

View Full Product Details