Product Details

SNP ID
rs55636450
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:118889090 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTACCACAGGCCAAGCACTTTGCA[C/T]CTCCTTACACCCCCCACCTCCCGCT
Phenotype
MIM: 609004 MIM: 601613
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BCL9L PubMed Links

Gene Details

Gene
BCL9L
Gene Name
B-cell CLL/lymphoma 9-like
There are no transcripts associated with this gene.

Gene
CXCR5
Gene Name
C-X-C motif chemokine receptor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001716.4 Intron NP_001707.1
NM_032966.2 Intron NP_116743.1
Gene
LOC101929133
Gene Name
uncharacterized LOC101929133
There are no transcripts associated with this gene.

View Full Product Details