Product Details

SNP ID
rs61749181
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67492106 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCAGGGTGGTGGGTGGTGTTCCC[A/G]GGCCCTCACGCTCCGCCTGGCTGGG
Phenotype
MIM: 605555 MIM: 608794
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AIP PubMed Links

Gene Details

Gene
AIP
Gene Name
aryl hydrocarbon receptor interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302959.1 3904 Intron NP_001289888.1
NM_001302960.1 3904 Intron NP_001289889.1
NM_003977.3 3904 Intron NP_003968.3
Gene
MIR6752
Gene Name
microRNA 6752
There are no transcripts associated with this gene.

Gene
PITPNM1
Gene Name
phosphatidylinositol transfer protein membrane associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130848.1 3904 Missense Mutation CCG,CTG P1220L NP_001124320.1
NM_004910.2 3904 Missense Mutation CCG,CTG P1221L NP_004901.2
XM_011545396.2 3904 Missense Mutation CCG,CTG P1220L XP_011543698.1
XM_017018586.1 3904 Missense Mutation CCG,CTG P1221L XP_016874075.1

View Full Product Details