Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001300942.1 | Intron | NP_001287871.1 | |||
NM_001300943.1 | Intron | NP_001287872.1 | |||
NM_001300944.1 | Intron | NP_001287873.1 | |||
NM_020193.4 | Intron | NP_064578.2 | |||
XM_005274112.4 | Intron | XP_005274169.1 | |||
XM_006718637.3 | Intron | XP_006718700.1 | |||
XM_006718638.3 | Intron | XP_006718701.1 | |||
XM_006718639.3 | Intron | XP_006718702.1 | |||
XM_011545173.2 | Intron | XP_011543475.1 | |||
XM_011545174.2 | Intron | XP_011543476.1 | |||
XM_011545175.2 | Intron | XP_011543477.1 | |||
XM_011545177.2 | Intron | XP_011543479.1 | |||
XM_011545178.2 | Intron | XP_011543480.1 | |||
XM_011545179.2 | Intron | XP_011543481.1 | |||
XM_017018021.1 | Intron | XP_016873510.1 | |||
XM_017018022.1 | Intron | XP_016873511.1 |