Product Details

SNP ID
rs58740322
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:100930041 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCTGAGGCTTAGATCTTCTCCTG[A/G]TTACTTGTCTGGAATGTGCTTGCTT
Phenotype
MIM: 610986
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRK1 PubMed Links

Gene Details

Gene
LRRK1
Gene Name
leucine rich repeat kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024652.4 Intron NP_078928.3
XM_005254979.3 Intron XP_005255036.1
XM_011522012.2 Intron XP_011520314.1
XM_011522013.2 Intron XP_011520315.1
XM_011522014.2 Intron XP_011520316.1
XM_011522015.2 Intron XP_011520317.1
XM_011522016.2 Intron XP_011520318.1
XM_011522017.2 Intron XP_011520319.1
XM_011522018.2 Intron XP_011520320.1
XM_011522019.2 Intron XP_011520321.1
XM_017022570.1 Intron XP_016878059.1

View Full Product Details