Product Details
- SNP ID
-
rs61157814
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:144984053 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAATTGGGGGGAAATACATAGTTCA[A/T]CAGCAGTTGTCAGAGACTTCAAACC
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF252P
PubMed Links
Gene Details
- Gene
- ZNF252P
- Gene Name
- zinc finger protein 252, pseudogene
There are no transcripts associated with this gene.
View Full Product Details