Product Details

SNP ID
rs61758748
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:590417 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGGACGGCGCGGCAGAGTCCCC[G/T]TACGCCTACAGTAACGGTAAGGCCC
Phenotype
MIM: 605754
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
LOC101929280 PubMed Links
Additional Information
For this assay, SNP(s) [rs183476305] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
LOC101929280
Gene Name
uncharacterized LOC101929280
There are no transcripts associated with this gene.

Gene
PIGQ
Gene Name
phosphatidylinositol glycan anchor biosynthesis class Q
There are no transcripts associated with this gene.

Gene
RAB40C
Gene Name
RAB40C, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172663.1 200 Silent Mutation CCG,CCT P42P NP_001166134.1
NM_001172664.1 200 Silent Mutation CCG,CCT P42P NP_001166135.1
NM_001172665.1 200 Silent Mutation CCG,CCT P42P NP_001166136.1
NM_001172666.1 200 Silent Mutation CCG,CCT P42P NP_001166137.1
NM_021168.4 200 Silent Mutation CCG,CCT P42P NP_066991.3

View Full Product Details