Product Details

SNP ID
rs60741147
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:1177338 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAATTTTACTGAATATTTTTATT[C/T]ACTTAACAGAAACATTCACTTATTA
Phenotype
MIM: 602065
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADARB2 PubMed Links

Gene Details

Gene
ADARB2
Gene Name
adenosine deaminase, RNA specific B2 (inactive)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018702.3 8401 UTR 3 NP_061172.1

View Full Product Details