Product Details
- SNP ID
-
rs56174221
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:22983352 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCCACCTCATCTTCTGGGGGATAA[G/A]CTTCAAGACCCCTGGTGGATGTCTG
- Phenotype
-
MIM: 606322
MIM: 608147
- Polymorphism
- G/A, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CYFIP1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11305791] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CYFIP1
- Gene Name
- cytoplasmic FMR1 interacting protein 1
There are no transcripts associated with this gene.
- Gene
- TUBGCP5
- Gene Name
- tubulin gamma complex associated protein 5
View Full Product Details