Product Details

SNP ID
rs71651693
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:154569506 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGAGCGGCTGGTGGAGCATCTC[C/T]TGGATCCTTCCCGCTACAACAAGCT
Phenotype
MIM: 118507
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHRNB2 PubMed Links

Gene Details

Gene
CHRNB2
Gene Name
cholinergic receptor nicotinic beta 2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000748.2 355 Silent Mutation CTG,TTG L37L NP_000739.1
XM_017000180.1 355 UTR 5 XP_016855669.1

View Full Product Details