Product Details

SNP ID
rs72761100
Assay Type
Functionally tested
NCBI dbSNP Submissions
25
Location
Chr.1:247961173 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTCAGAAACACAGAAGAATCAG[G/T]CTTGGTATACACACATTATGCACAA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR2AK2 PubMed Links

Gene Details

Gene
OR2AK2
Gene Name
olfactory receptor family 2 subfamily AK member 2
There are no transcripts associated with this gene.

Gene
OR2L13
Gene Name
olfactory receptor family 2 subfamily L member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304535.1 Intron NP_001291464.1
NM_175911.3 Intron NP_787107.1
XM_011544169.2 Intron XP_011542471.1

View Full Product Details