Product Details

SNP ID
rs73019262
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:154701001 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCATATATAATTAATTTCAATGA[A/G]CTGCATACAATTTTTTAAAATTTAA
Phenotype
MIM: 601534
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNJ3 PubMed Links

Gene Details

Gene
KCNJ3
Gene Name
potassium voltage-gated channel subfamily J member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001260508.1 Intron NP_001247437.1
NM_001260509.1 Intron NP_001247438.1
NM_001260510.1 Intron NP_001247439.1
NM_002239.3 Intron NP_002230.1

View Full Product Details