Product Details

SNP ID
rs73968433
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:157413560 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCAAACTAAAGTGATGCTTGCTCC[G/T]TCCCTTCCCTGCCCCTTTTTTGCAT
Phenotype
MIM: 604448
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CYTIP PubMed Links

Gene Details

Gene
CYTIP
Gene Name
cytohesin 1 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004288.4 3460 Intron NP_004279.3
XM_017005386.1 3460 UTR 3 XP_016860875.1

View Full Product Details