Product Details

SNP ID
rs73167497
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:153488053 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCTGAATAGAACCTCAAACCACA[A/G]AGGATTTGCAGTAATGACTCCATCA
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
C3orf79 PubMed Links

Gene Details

Gene
C3orf79
Gene Name
chromosome 3 open reading frame 79
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101337.1 Intron NP_001094807.1
Gene
LOC105374165
Gene Name
uncharacterized LOC105374165
There are no transcripts associated with this gene.

View Full Product Details