Product Details

SNP ID
rs73178410
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:169913772 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACAATTTGTCTACATGTGTCTAAA[A/G]TAGAAAGGGGTGCCCTGAAAATATG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SAMD7 PubMed Links

Gene Details

Gene
SAMD7
Gene Name
sterile alpha motif domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304366.1 Intron NP_001291295.1
NM_182610.3 Intron NP_872416.1

View Full Product Details