Product Details

SNP ID
rs73883571
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:179607081 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGATAAACTGCAGCAAAATTTCAC[C/T]CCCAGTTTCTCTTTCCACATTATTG
Phenotype
MIM: 611852 MIM: 603841
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRPL47 PubMed Links

Gene Details

Gene
MRPL47
Gene Name
mitochondrial ribosomal protein L47
There are no transcripts associated with this gene.

Gene
NDUFB5
Gene Name
NADH:ubiquinone oxidoreductase subunit B5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199957.1 Intron NP_001186886.1
NM_001199958.1 Intron NP_001186887.1
NM_002492.3 Intron NP_002483.1

View Full Product Details