Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001014809.2 | Intron | NP_001014809.1 | |||
NM_001288661.1 | Intron | NP_001275590.1 | |||
NM_001288662.1 | Intron | NP_001275591.1 | |||
NM_001313.4 | Intron | NP_001304.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001306090.1 | Intron | NP_001293019.1 | |||
NM_001306092.1 | Intron | NP_001293021.1 | |||
NM_153717.2 | Intron | NP_714928.1 | |||
XM_006713865.3 | Intron | XP_006713928.1 | |||
XM_006713866.3 | Intron | XP_006713929.1 | |||
XM_011513419.2 | Intron | XP_011511721.1 | |||
XM_017007883.1 | Intron | XP_016863372.1 |