Product Details

SNP ID
rs73195118
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:3467758 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCAGACGACAGCCAGTCACTGCT[A/C]ACGGTTCTGGGGGCTGGAGGTCCAA
Phenotype
MIM: 610285
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DOK7 PubMed Links

Gene Details

Gene
DOK7
Gene Name
docking protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164673.1 Intron NP_001158145.1
NM_001256896.1 Intron NP_001243825.1
NM_001301071.1 Intron NP_001288000.1
NM_173660.4 Intron NP_775931.3
XM_011513435.2 Intron XP_011511737.1
XM_011513437.2 Intron XP_011511739.1
XM_017008038.1 Intron XP_016863527.1

View Full Product Details