Product Details

SNP ID
rs528920
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:167785227 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATAGAAAACTATACAGCCACTAA[C/T]ACAAATGCGATTTTGAAAAACATTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC01558 PubMed Links

Gene Details

Gene
LINC01558
Gene Name
long intergenic non-protein coding RNA 1558
There are no transcripts associated with this gene.

Gene
LOC441179
Gene Name
uncharacterized LOC441179
There are no transcripts associated with this gene.

View Full Product Details