Product Details

SNP ID
rs72841516
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:26370967 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAATCAGAAGGAGATTCTAAAAA[C/T]CTAGTAAAAAGTTTTGGCCAGGCAT
Phenotype
MIM: 613594
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BTN3A2 PubMed Links

Gene Details

Gene
BTN3A2
Gene Name
butyrophilin subfamily 3 member A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197246.2 Intron NP_001184175.1
NM_001197247.2 Intron NP_001184176.1
NM_001197248.2 Intron NP_001184177.1
NM_001197249.2 Intron NP_001184178.1
NM_007047.4 Intron NP_008978.2
XM_005248827.3 Intron XP_005248884.1
XM_005248831.3 Intron XP_005248888.1
XM_005248832.3 Intron XP_005248889.1
XM_006714979.3 Intron XP_006715042.1
XM_006714980.3 Intron XP_006715043.1
XM_006714981.3 Intron XP_006715044.1
XM_006714982.3 Intron XP_006715045.1
XM_011514267.2 Intron XP_011512569.1
XM_011514268.2 Intron XP_011512570.1
XM_011514269.2 Intron XP_011512571.1
XM_011514270.2 Intron XP_011512572.1
XM_011514271.2 Intron XP_011512573.1
XM_017010211.1 Intron XP_016865700.1
XM_017010212.1 Intron XP_016865701.1
XM_017010213.1 Intron XP_016865702.1
XM_017010214.1 Intron XP_016865703.1
XM_017010215.1 Intron XP_016865704.1
XM_017010216.1 Intron XP_016865705.1
XM_017010217.1 Intron XP_016865706.1

View Full Product Details