Product Details

SNP ID
rs73740918
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:53066404 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAAATGAGAGACTGAGCTAAGTT[A/G]CAGAGCTTGACAGTGCAGATTCCAT
Phenotype
MIM: 609091 MIM: 612325
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBXO9 PubMed Links

Gene Details

Gene
FBXO9
Gene Name
F-box protein 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012347.4 Intron NP_036479.1
NM_033480.2 Intron NP_258441.1
NM_033481.3 Intron NP_258442.2
XM_005248995.4 Intron XP_005249052.1
XM_005248996.4 Intron XP_005249053.1
Gene
ICK
Gene Name
intestinal cell (MAK-like) kinase
There are no transcripts associated with this gene.

Gene
LOC105375092
Gene Name
uncharacterized LOC105375092
There are no transcripts associated with this gene.

View Full Product Details