Product Details

SNP ID
rs72556347
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:18132136 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCATAAGTCCACAAACTTACCAA[G/A]CAACCTTTCAATTTCAGCATGTGGA
Phenotype
MIM: 608072 MIM: 187680
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
NHLRC1 PubMed Links

Gene Details

Gene
NHLRC1
Gene Name
NHL repeat containing E3 ubiquitin protein ligase 1
There are no transcripts associated with this gene.

Gene
TPMT
Gene Name
thiopurine S-methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000367.3 1093 Missense Mutation CTT,TTT L208F NP_000358.1
XM_011514839.2 1093 Intron XP_011513141.1
XM_017011241.1 1093 Missense Mutation CTT,TTT L208F XP_016866730.1

View Full Product Details