Product Details

SNP ID
rs73136380
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:66131301 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTAACCTAATTTTTATTCTGTTT[A/G]TAGAAGTGAGAGGAGTTTGGTCAAG
Phenotype
MIM: 606121
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRCP PubMed Links

Gene Details

Gene
CRCP
Gene Name
CGRP receptor component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040647.1 Intron NP_001035737.1
NM_001040648.1 Intron NP_001035738.1
NM_001142414.1 Intron NP_001135886.1
NM_014478.4 Intron NP_055293.1

View Full Product Details