Product Details

SNP ID
rs73403251
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:99503706 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAAGCACCATATGAAAAGGAGAG[A/C]ACTGGGAGGTCGATGCGGGCGCATC
Phenotype
MIM: 611272
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZKSCAN5 PubMed Links

Gene Details

Gene
ZKSCAN5
Gene Name
zinc finger with KRAB and SCAN domains 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318082.1 Intron NP_001305011.1
NM_001318083.1 Intron NP_001305012.1
NM_001318084.1 Intron NP_001305013.1
NM_014569.3 Intron NP_055384.1
NM_145102.3 Intron NP_659570.1
XM_011515999.2 Intron XP_011514301.1
XM_017011918.1 Intron XP_016867407.1
XM_017011919.1 Intron XP_016867408.1
XM_017011920.1 Intron XP_016867409.1
XM_017011921.1 Intron XP_016867410.1
XM_017011922.1 Intron XP_016867411.1
XM_017011923.1 Intron XP_016867412.1
XM_017011924.1 Intron XP_016867413.1
XM_017011925.1 Intron XP_016867414.1
XM_017011926.1 Intron XP_016867415.1
XM_017011927.1 Intron XP_016867416.1
Gene
ZNF394
Gene Name
zinc finger protein 394
There are no transcripts associated with this gene.

View Full Product Details