Product Details

SNP ID
rs73713803
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:101169997 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGGGGATCGCAGTCTCCGCCAAT[A/C]CAGAGCAGGCTTCGCGGGGGCGGTG
Phenotype
MIM: 603531 MIM: 615783 MIM: 602186
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
AP1S1 PubMed Links

Gene Details

Gene
AP1S1
Gene Name
adaptor related protein complex 1 sigma 1 subunit
There are no transcripts associated with this gene.

Gene
NAT16
Gene Name
N-acetyltransferase 16 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198571.2 Intron NP_940973.2
XM_005250325.1 Intron XP_005250382.1
XM_011516168.1 Intron XP_011514470.1
Gene
VGF
Gene Name
VGF nerve growth factor inducible
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003378.3 Intron NP_003369.2
XM_005250561.4 Intron XP_005250618.1
XM_011516549.2 Intron XP_011514851.1
XM_017012579.1 Intron XP_016868068.1

View Full Product Details