Product Details

SNP ID
rs67675731
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:138802745 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCGCTTTGTGAGGAAGGCATAAGC[C/T]AACGTGTAAAAGCACGCAGCACAGT
Phenotype
MIM: 605239
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP6V0A4 PubMed Links

Gene Details

Gene
ATP6V0A4
Gene Name
ATPase H+ transporting V0 subunit a4
There are no transcripts associated with this gene.

Gene
TMEM213
Gene Name
transmembrane protein 213
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085429.1 Intron NP_001078898.1
XM_005250167.3 Intron XP_005250224.1

View Full Product Details