Product Details

SNP ID
rs73260945
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:80629854 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTTCACTTCATGCAGTTTTGTTTG[C/T]TTGTTAATCCAGTGTTACTAGAACA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF704 PubMed Links

Gene Details

Gene
ZNF704
Gene Name
zinc finger protein 704
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033723.2 13614 UTR 3 NP_001028895.1
XM_005251279.4 13614 UTR 3 XP_005251336.2
XM_017013725.1 13614 UTR 3 XP_016869214.1

View Full Product Details