Product Details
- SNP ID
-
rs73389479
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:4994988 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAACTAAAGATTAAGTAGAAGAGGA[A/G]TTGCTGATTCAAAAGTGGTATATGT
- Phenotype
-
MIM: 606414
MIM: 147796
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
INSL6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs540777357] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- INSL6
- Gene Name
- insulin like 6
- Gene
- JAK2
- Gene Name
- Janus kinase 2
View Full Product Details