Product Details

SNP ID
rs73436358
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:26670041 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGTCAACTCGCCTTTTAATCTGC[A/G]ATACCCTCATTCTCCAGAACTTTAA
Phenotype
MIM: 610110 MIM: 612455
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANO3 PubMed Links

Gene Details

Gene
ANO3
Gene Name
anoctamin 3
There are no transcripts associated with this gene.

Gene
SLC5A12
Gene Name
solute carrier family 5 member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178498.3 3157 UTR 3 NP_848593.2
XM_006718155.3 3157 UTR 3 XP_006718218.1
XM_006718156.3 3157 Intron XP_006718219.1
XM_011519920.2 3157 UTR 3 XP_011518222.1
XM_011519921.2 3157 Intron XP_011518223.1
XM_017017244.1 3157 UTR 3 XP_016872733.1

View Full Product Details