Product Details

SNP ID
rs72885364
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:40111892 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGTCAAGTCTGGTTTATTTAAA[A/C]CTTGCATCAAATTTATTTCCCAAAA
Phenotype
MIM: 608817
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LRRC4C PubMed Links

Gene Details

Gene
LRRC4C
Gene Name
leucine rich repeat containing 4C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258419.1 5625 Intron NP_001245348.1
NM_020929.2 5625 Intron NP_065980.1
XM_011520238.2 5625 UTR 3 XP_011518540.1
XM_011520239.2 5625 UTR 3 XP_011518541.1
XM_011520240.2 5625 UTR 3 XP_011518542.1
XM_011520241.2 5625 UTR 3 XP_011518543.1
XM_011520242.2 5625 UTR 3 XP_011518544.1
XM_011520243.2 5625 UTR 3 XP_011518545.1
XM_011520244.2 5625 UTR 3 XP_011518546.1
XM_017018070.1 5625 UTR 3 XP_016873559.1
XM_017018071.1 5625 UTR 3 XP_016873560.1
XM_017018072.1 5625 UTR 3 XP_016873561.1
XM_017018073.1 5625 UTR 3 XP_016873562.1
XM_017018074.1 5625 UTR 3 XP_016873563.1
XM_017018075.1 5625 UTR 3 XP_016873564.1
XM_017018076.1 5625 UTR 3 XP_016873565.1
XM_017018077.1 5625 UTR 3 XP_016873566.1
XM_017018078.1 5625 UTR 3 XP_016873567.1
XM_017018079.1 5625 UTR 3 XP_016873568.1

View Full Product Details