Product Details

SNP ID
rs73400692
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5233124 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGAGGTAGGCAGATACATGCAT[A/G]TGGTTAACAGAGAAATAAAGACTGG
Phenotype
MIM: 141900 MIM: 142000
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HBB PubMed Links

Gene Details

Gene
HBB
Gene Name
hemoglobin subunit beta
There are no transcripts associated with this gene.

Gene
HBBP1
Gene Name
hemoglobin subunit beta pseudogene 1
There are no transcripts associated with this gene.

Gene
HBD
Gene Name
hemoglobin subunit delta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000519.3 Intron NP_000510.1

View Full Product Details