Product Details

SNP ID
rs73040581
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:2900491 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGTCTATGAAATTACAAAAGTAA[A/G]GCCTAAGGGAGATGGCAAGCTTTCC
Phenotype
MIM: 604730
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TULP3 PubMed Links

Gene Details

Gene
TULP3
Gene Name
tubby like protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160408.1 Intron NP_001153880.1
NM_003324.4 Intron NP_003315.2

View Full Product Details