Product Details

SNP ID
rs72646483
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:23183137 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCTTCCCTGCCGGGGTGAGTCG[A/C]GTGGACCTGTCCGCTACTGTTCCTT
Phenotype
MIM: 600761
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SCNN1G PubMed Links

Gene Details

Gene
SCNN1G
Gene Name
sodium channel epithelial 1 gamma subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039.3 Intron NP_001030.2

View Full Product Details